Key Documents
SYNV130711
Synthware conical bottom reaction vial with holed compression cap
1 mL, joint: ST/NS 7/10
Synonym(s):
Synthware V130711, reaction tube
Select a Size
1 EA
€19.90
€19.90
Estimated to ship on31 March 2025
Select a Size
Change View
1 EA
€19.90
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
€19.90
Estimated to ship on31 March 2025
Joint
joint: ST/NS7/10
packaging
pkg of 1 ea
manufacturer/tradename
Synthware V130711
capacity
1 mL
General description
Supplied with one holed compression cap, one Viton® O-ring and one PTFE -faced silicone rubber septum. For replacement caps, see RC112; For Viton® O-rings, see R014; for septa, see R1420.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)