Key Documents
SYNS734040
Synthware sublimator, complete
40 mL, complete
Synonym(s):
Synthware S734040, sublimation apparatus
Select a Size
1 EA
€314.00
€314.00
Check Cart for Availability
Select a Size
Change View
1 EA
€314.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
€314.00
Check Cart for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S734040
capacity
40 mL
General description
A grease-free, two-piece sublimation apparatus consisting of an upper water cooled condenser and lower vessel to hold the material to be sublimed. The sublimate collects on the lower end of the condenser which has a flat bottom. Condenser is constructed using a grease-free, threaded valve. Connection of the condenser and lower vessel via an O-ring joint enables the two parts to be gently separated for maximum product recovery. Complete apparatus consists of the cold finger condenser, lower vessel, Viton® O-ring and stainless steel pinch clamp.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)