Key Documents
SYNS532440
Synthware PTFE stopper with extraction nut
joint: ST/NS 24/40
Synonym(s):
Synthware S532440
Select a Size
1 EA
€43.60
€43.60
Check Cart for Availability
Select a Size
Change View
1 EA
€43.60
About This Item
UNSPSC Code:
41122500
NACRES:
NB.51
€43.60
Check Cart for Availability
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S532440
General description
PTFE stopper has a Nylon extraction nut permitting easy removal of the stopper from the glass outer joint.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)