Documents
SYNM211042
Synthware nitrogen/argon line manifold with PTFE stopcocks
port size 4, Hose Connections: left-right, stopcock size 2 mm
Synonym(s):
Synthware M211042, vacuum manifold, vacuum unit
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M211042
port size
4
stopcock size
2 mm
Related Categories
General description
With 14/20 outer joint at the center to fit a rubber septum for keeping syringes under an inert environment cutting down on the number used. Main tube is 22 mm O.D. with both ends connections(10 mm). Delivery ports have 8 mm hose connections.
Legal Information
Synthware is a trademark of Kemtech America Inc.