Key Documents
SYNF411125
Synthware two neck round bottom flask with angled side neck
25 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Synonym(s):
Synthware F411125, 2-neck round flask
Select a Size
1 EA
€49.40
€49.40
Check Cart for Availability
Select a Size
Change View
1 EA
€49.40
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
€49.40
Check Cart for Availability
material
borosilicate glass flask
round bottom
two-neck (angled side neck)
Joint
center joint: ST/NS14/20
side joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F411125
capacity
25 mL
25 mL
General description
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)