Key Documents
SYNF318100
Synthware recovery flask
100 mL, joint: ST/NS 29/42
Synonym(s):
Synthware F318100, oval shape flask
Select a Size
1 EA
€28.40
€28.40
Estimated to ship on30 April 2025
Select a Size
Change View
1 EA
€28.40
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
€28.40
Estimated to ship on30 April 2025
material
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS29/42
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F318100
capacity
100 mL
100 mL
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)