Skip to Content
Merck
All Photos(1)

Documents

SYNF198100

Synthware powder funnel with flattened side

joint: ST/NS 29/20, top diam. 100 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.51

Joint

joint: ST/NS29/20

packaging

pkg of 1 ea

manufacturer/tradename

Synthware F198100

top diam.

100 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Funnels have 60° angled side with a standard taper inner joint.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service