Skip to Content
Merck

Skip To

SYNC509300

Synthware™ Liebig condenser with 8 mm OD removable hose connections

joint: ST/NS 19/22, jacket L 300 mm

Synonym(s):

Synthware C509300, straight condenser

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
€ 117,00

About This Item

NACRES:
NB.51
UNSPSC Code:
41121800

€ 117,00


Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


joint

joint: ST/NS19/22

packaging

pkg of 1 ea

manufacturer/tradename

Synthware C509300

jacket L

300 mm

General description

Synthware™ Liebig condenser features a 300 mm jacketed length for superior cooling efficiency. With standard taper outer joint at the top (19/22) and a lower inner drip tip joint. Made from durable borosilicate glass.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SYNC501120SYNC504250SYNC509180
description

joint: ST/NS 19/22, jacket L 300 mm

description

joint: ST/NS 14/20, jacket L 120 mm

description

joint: ST/NS 24/40, jacket L 250 mm

description

joint: ST/NS 19/22, jacket L 180 mm

packaging

pkg of 1 ea

packaging

pkg of 1 ea

packaging

pkg of 1 ea

packaging

pkg of 1 ea

joint

joint: ST/NS19/22

joint

joint: ST/NS14/20

joint

joint: ST/NS24/40

joint

joint: ST/NS19/22

manufacturer/tradename

Synthware C509300

manufacturer/tradename

Synthware C501120

manufacturer/tradename

Synthware C504250

manufacturer/tradename

Synthware C509180

jacket L

300 mm

jacket L

120 mm

jacket L

250 mm

jacket L

180 mm


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library