As listed on the product detail page under the title of this product, the inner diameter is 12.4 mm.
SYNC361720
Synthware™ chromatography column with PTFE stopcock
I.D. 13.4 mm, L 203 mm
Synonym(s):
Synthware C361720, glass chromatograpy column
Select a Size
€87.40
Select a Size
About This Item
€87.40
Recommended Products
grade
for chromatography
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C361720
I.D.
13.4 mm
L
203 mm
suitability
suitable for chromatography
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the diameter of this SYNC361720 column?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service