Key Documents
SYNA851420
Synthware U-shaped drying tube
joint: ST/NS 14/20
Synonym(s):
Synthware A851420, Drying tube 90° bent
Select a Size
1 EA
€35.50
€35.50
Check Cart for Availability
Select a Size
Change View
1 EA
€35.50
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
€35.50
Check Cart for Availability
material
colorless tube
glass tube
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A851420
General description
With joint. Cotton is usually used to contain drying agent.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)