Key Documents
SYNA254024
Synthware enlarging adapter
top joint: ST/NS 40/50, bottom joint: ST/NS 24/40
Synonym(s):
Synthware A254024, glass connecting adapter
Select a Size
1 EA
€61.50
€61.50
Check Cart for Availability
Select a Size
Change View
1 EA
€61.50
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
€61.50
Check Cart for Availability
Joint
bottom joint: ST/NS24/40
joint: ST/NS top40/50
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A254024
General description
Synthware enlarging adapter is used for connecting dissimilar standard taper joint sizes. The top joint (40/50) is larger that the bottom joint (24/40).
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)