Key Documents
SYNA201024
Synthware reducing adapter
female joint: ST/NS 10/30, male joint: ST/NS 24/40
Synonym(s):
Synthware A201024, glass reducing adapter, connecting adapter
Select a Size
1 EA
€29.90
€29.90
Check Cart for Availability
Select a Size
Change View
1 EA
€29.90
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
€29.90
Check Cart for Availability
Joint
female joint: ST/NS10/30
male joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A201024
Related Categories
General description
Synthware reducing adapter is used for connecting dissimilar standard taper joint sizes. The bottom joint (24/40) is larger than the top joint (10/30).
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)