Skip to Content
Merck
All Photos(1)

Key Documents

SLW718347R

Pyrex® Pestle

glazed finish, L 130 mm

Synonym(s):

Pyrex, SciLabware 718347R

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€22.10

€22.10


Check Cart for Availability


Select a Size

Change View
1 EA
€22.10

About This Item

UNSPSC Code:
41121800
NACRES:
NB.35

€22.10


Check Cart for Availability

material

borosilicate glass

manufacturer/tradename

718347R
SciLabware

L

130 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • Ideal for crushing solids into powders for easier handling
  • Manufactured from Pyrex borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • Available in a rough or glazed finish
  • For use with Pyrex Mortars

Features and Benefits

  • Manufactured from borosilicate glass 3.3
  • Available in a rough or glazed finish
  • Ideal for crushing solids into powders for easier handling
  • For use with PYREX® Mortars
  • Pestles and mortars should be ordered separately.

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service