SLW1568/04M
Pyrex® Stopcocks, with Rotaflo® PTFE key
bore 3 mm , key
Synonym(s):
Pyrex, SciLabware 1568/04M
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Joint
cone joint 29/32
manufacturer/tradename
1568/04M
SciLabware
L
53 mm
bore
3 mm , key
Looking for similar products? Visit Product Comparison Guide
General description
- For use with Aspirator bottles SLW1541: and SLW1569:
- Type 1568/04M with Rotaflo PTFE key which is chemically inert and requires no lubrication
Features and Benefits
• For use with Aspirator bottles 1541 and 1569• Type 1568/04M with Rotaflo® PTFE key which is chemically inert and requires no lubrication• Type 1571/02M with traditional glass key
Legal Information
PYREX is a registered trademark of Corning, Inc.
Rotaflo is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service