Skip to Content
Merck
All Photos(1)

Key Documents

SIK7706-10

Sigma-Aldrich

1-Methyl-3-(dodecylphosphonic acid)imidazolium bromide

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€530.00

€530.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 G
€530.00

About This Item

Empirical Formula (Hill Notation):
C16H32BrN2O3P
Molecular Weight:
411.31
UNSPSC Code:
12352103
NACRES:
NA.23

€530.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Assay

≥95%

form

solid

shelf life

3 years under the recommended conditions

color

white

Application

This coupling agent is used to create positively charged surfaces and potential antimicrobial surfaces.

Legal Information

Product of SiKEMIA

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service