Skip to Content
Merck
All Photos(1)

Documents

SIK2801-10

Sigma-Aldrich

4,4′-Sulfonyldibenzoic acid

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O6S
CAS Number:
Molecular Weight:
306.29
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

≥97%

InChI

1S/C14H10O6S/c15-13(16)9-1-5-11(6-2-9)21(19,20)12-7-3-10(4-8-12)14(17)18/h1-8H,(H,15,16)(H,17,18)

InChI key

SQJQLYOMPSJVQS-UHFFFAOYSA-N

Related Categories

Legal Information

Product of SiKEMIA

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service