Skip to Content
Merck
All Photos(1)

Key Documents

QBD10869

Sigma-Aldrich

Amino-dPEG®36-OH

>95% (HPLC)

Synonym(s):

Amino-PEG36-OH, Hydroxy-PEG-amine, Hydroxy-PEG36-amine, OH-PEG36-NH2, PEG1500-amine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€495.00

€495.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€495.00

About This Item

Empirical Formula (Hill Notation):
C72H147NO36
Molecular Weight:
1602.92
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

€495.00


Check Cart for Availability

Request a Bulk Order

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI key

MAESJFAEMPPQIN-UHFFFAOYSA-N

Features and Benefits

Amino-dPEG36-OH, also known as Amino-dPEG36-alcohol, is a very long (109 atoms, 127.7 Å) monodispersed PEGylation reagent functionalized with a primary amine on one end of the molecule that reacts with carboxylic acids, aldehydes, and ketones. Reacting the amine group with carboxylic acids yields stable amides while reacting the amine group with aldehydes and ketones forms Schiff bases that are reducible to secondary amines for stability. If needed, various functional and reactive groups can modify the terminal alcohol on the opposite end of the molecule to provide the dPEG product with additional reactivity or crosslinking capability. dPEG36 is a monodisperse equivalent of the polymeric PEG1500. Amino-dPEG36-OH can be used for passivation of carboxylate-functionalized surfaces. The non-immunogenic terminal alcohol of the PEG chain imparts hydrophilicity to the conjugate molecule or surface. Consequently, conjugate molecules are water-soluble.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service