Documents
PL800GB-9-10
Plas-Labs butyl glove box gloves
for 1 piece glove set, size 9 (for 10 in. ports)
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
material
butyl rubber
size
9 (for 10 in. ports)
feature
reusable
packaging
pack of 1 pair
manufacturer/tradename
Plas-Labs 800GB-9-10
General description
Plas-Labs black butyl rubber gloves for highest impermeability to water vapor, gases, and toxic fluids
Note: Order sleeves, gloves, and glove over sleeve connectors separately.