All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H5NO3S
CAS Number:
Molecular Weight:
159.16
Beilstein:
120822
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
98%
mp
>300 °C (lit.)
SMILES string
OS(=O)(=O)c1cccnc1
InChI
1S/C5H5NO3S/c7-10(8,9)5-2-1-3-6-4-5/h1-4H,(H,7,8,9)
InChI key
DVECLMOWYVDJRM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service