Skip to Content
Merck
All Photos(3)

Key Documents

P28808

Sigma-Aldrich

Phenylphosphinic acid

99%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€42.00
500 G
€134.00

€42.00


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
100 G
€42.00
500 G
€134.00

About This Item

Linear Formula:
C6H5P(O)(OH)H
CAS Number:
Molecular Weight:
142.09
Beilstein:
2802244
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€42.00


Only 0 left in stock

Request a Bulk Order

Quality Level

Assay

99%

form

powder

mp

83-85 °C (lit.)

SMILES string

O[PH](=O)c1ccccc1

InChI

1S/C6H7O2P/c7-9(8)6-4-2-1-3-5-6/h1-5,9H,(H,7,8)

InChI key

MLCHBQKMVKNBOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenylphosphinic acid is also known as phenylphosphonous acid. Kinetics of oxidation of phenylphosphinic acid by metal and non metal oxidants was investigated in a study.2

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Gervais et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 187(1), 131-140 (2007-05-08)
The complete set of NMR parameters for (17)O enriched phenylphosphinic acid C(6)H(5)HP( *)O(*OH) is calculated from first principles by using the Gauge Including Projected Augmented Wave (GIPAW) approach [C.J. Pickard, F. Mauri, All-electron magnetic response with pseudopotentials: NMR chemical shifts
Loïc J Charbonnière et al.
Inorganic chemistry, 44(20), 7151-7160 (2005-09-27)
A new ligand, LC, bis-[(6'-carboxy-2,2'-bipyridine-6-yl)]phenylphosphine oxide, in which the tridentate 6-carboxy-2,2'-bipyridyl arms are directly linked to a phenylphosphine oxide fragment, has been synthesized. The corresponding [Ln.LC]Cl.xH2O complexes (Ln = Eu, x = 4, and Tb, x = 3) were isolated
E Maria Donner et al.
Drug and chemical toxicology, 26(2), 125-133 (2003-06-21)
Phenylphosphinic acid was fed in graded concentrations of either 0 (control), 100, 1,000, or 10,000 ppm to rats for 28 days. These concentrations provided average daily doses of 8, 76, and 779 mg/kg (males) and 9, 83, and 859 mg/kg
Kunio Ikemura et al.
Dental materials journal, 28(3), 267-276 (2009-08-11)
The behavior of water-soluble photoinitiators with crown ethers in dental adhesives is unknown. This study investigated the effect of sodium acylphosphine oxide (APO-Na) with crown ether in a hydrophobic adhesive on adhesion to teeth. Sodium 2,4,6-trimethylbenzoyl-phenylphosphine oxide (TMPO-Na = APO-Na)
Li-Biao Han et al.
The Journal of organic chemistry, 70(24), 10121-10123 (2005-11-19)
[reaction: see text] A variety of functionalized optically pure (R(P))-alkylphenylphosphinates are readily prepared by stereospecific radical or base-catalyzed additions of the easily available (R(P))-menthyl phenylphosphinate to alkenes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service