Skip to Content
Merck
All Photos(1)

Documents

P2880

Sigma-Aldrich

Propionylthiocholine iodide

≥98%

Synonym(s):

(2-Mercaptoethyl)trimethylammonium iodide propionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H18NOSI
CAS Number:
Molecular Weight:
303.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

mp

200-202 °C (lit.)

storage temp.

−20°C

SMILES string

[I-].CCC(=O)SCC[N+](C)(C)C

InChI

1S/C8H18NOS.HI/c1-5-8(10)11-7-6-9(2,3)4;/h5-7H2,1-4H3;1H/q+1;/p-1

InChI key

BJQIXLJDWWVGAE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia G Rodrigues et al.
American journal of physiology. Heart and circulatory physiology, 316(3), H459-H475 (2018-12-12)
Several studies have demonstrated that administration of doxorubicin (DOXO) results in cardiotoxicity, which eventually progresses to dilated cardiomyopathy. The present work aimed to evaluate the early myocardial changes of DOXO-induced cardiotoxicity. Male New Zealand White rabbits were injected intravenously with
J M Monserrat et al.
Comparative biochemistry and physiology. Part C, Pharmacology, toxicology & endocrinology, 120(2), 193-199 (1998-11-25)
In this study, some kinetic and toxicological parameters of the thoracic ganglia cholinesterase from the estuarine crab Chasmagnathus granulata were determined. Effects of the type and concentration of substrate, pH (6.80-8.50), incubation temperature (5-35 degrees C) and eserine on the
R N Guedes et al.
Comparative biochemistry and physiology. Part C, Pharmacology, toxicology & endocrinology, 119(2), 205-210 (1998-07-21)
Acetylcholinesterase (AChE, EC 3.1.1.7) purified from the lesser grain borer (Rhyzopertha dominica) was significantly inhibited by higher concentrations of the substrates acetylthiocholine (ATC), acetyl-(beta-methyl) thiocholine (A beta MTC) and propionylthiocholine (PTC). 2. The efficiency of AChE for hydrolyzing different substrates
G Albendín et al.
Fish physiology and biochemistry, 43(2), 455-464 (2016-10-08)
The characterization of cholinesterase activity in brain and muscle of gilthead seabream was carried out using four specific substrates and three selective inhibitors. In addition, K
Magda A Mohamed et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 146(3), 314-324 (2007-05-29)
Acetylcholinesterases (AChEs) have been estimated in the infective juveniles (IJs) of eight different strains of heterorhabditid nematodes. The enzyme content ranged from 45.6 to 421.3 units/10(5) IJs with specific activity 34.0 to 82.6 units/mg protein. The isoenzyme patterns revealed the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service