Skip to Content
Merck
All Photos(1)

Key Documents

P25701

Sigma-Aldrich

Hexylbenzene

97%

Synonym(s):

1-Phenylhexane

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€81.60
100 G
€190.00

€81.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€81.60
100 G
€190.00

About This Item

Linear Formula:
C6H5(CH2)5CH3
CAS Number:
Molecular Weight:
162.27
Beilstein:
1904786
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€81.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.486 (lit.)

bp

226 °C (lit.)

mp

−61 °C (lit.)

density

0.861 g/mL at 25 °C (lit.)

SMILES string

CCCCCCc1ccccc1

InChI

1S/C12H18/c1-2-3-4-6-9-12-10-7-5-8-11-12/h5,7-8,10-11H,2-4,6,9H2,1H3

InChI key

LTEQMZWBSYACLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik A Rodríguez-Morales et al.
Chemosphere, 171, 435-445 (2016-12-30)
The environmental bioavailability of zinc (II), i.e., the uptake of the element by an organism, was determined using two microalgae species, Scenedesmus acutus and Pseudokirchneriella subcapitata, and estimated using hollow fiber supported liquid membrane (HF-SLM) device as the chemical surrogate.
Iria Rodriguez-Escontrela et al.
Journal of colloid and interface science, 504, 404-416 (2017-06-09)
Chemical flooding with surfactants for reducing oil-brine interfacial tensions (IFTs) to mobilize residual oil trapped by capillary forces has a great potential for Enhanced Oil Recovery (EOR). Surface-active ionic liquids (SAILs) constitute a class of surfactants that has recently been
Hiroshi Kobayashi et al.
Journal of chromatography. A, 1109(1), 2-9 (2005-12-20)
The contribution of molecular diffusion to peak broadening was studied in a reversed-phase HPLC system, consisting of a monolithic silica C18 column and methanol-water mobile phase. Study on the band broadening effect of holding a solute in a column or
Metabolism of n-butyl, n-hexyl, and n-octyl benzene in the bile of rainbow trout.
J Hellou et al.
Bulletin of environmental contamination and toxicology, 39(2), 182-187 (1987-08-01)
Yi Min et al.
Journal of chromatography. A, 1356, 148-156 (2014-07-08)
In this work, 1.9 μm reversed-phase packing materials with superficially porous structure were prepared to achieve the rapid and high efficient separation of peptides and proteins. The silica particles were synthesized via three steps, nonporous silica particle preparation by a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service