Skip to Content
Merck
All Photos(1)

Key Documents

P209

Sigma-Aldrich

Parabanic acid

99%

Synonym(s):

Imidazolidinetrione, Oxalylurea, Trioxoimidazolidine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€172.00

€172.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€172.00

About This Item

Empirical Formula (Hill Notation):
C3H2N2O3
CAS Number:
Molecular Weight:
114.06
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39161001
PubChem Substance ID:
NACRES:
NA.22

€172.00


Estimated to ship on26 May 2025


Request a Bulk Order

Assay

99%

form

powder

mp

249 °C (dec.) (lit.)

SMILES string

O=C1NC(=O)C(=O)N1

InChI

1S/C3H2N2O3/c6-1-2(7)5-3(8)4-1/h(H2,4,5,6,7,8)

InChI key

ZFLIKDUSUDBGCD-UHFFFAOYSA-N

Application

Reactant for:
  • Enantiospecific assembly of homochiral, hexanuclear palladium complexes
  • Mitsunobu reactions
  • Quantitative cascade condensation reactions

Reactant for synthesis of:
  • Pyridine derivatives
  • Nitroesters
  • Parabanic acid derivatives

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduction of oxalogenesis in a rapid gas chromatographic procedure for the analysis of oxalate ion in urine.
H A Moye et al.
Clinica chimica acta; international journal of clinical chemistry, 129(3), 385-390 (1983-04-25)
X M He et al.
Acta crystallographica. Section B, Structural science, 44 ( Pt 3), 271-281 (1988-06-01)
The crystal structure of parabanic acid (1H,3H-imidazoletrione, C3H2N2O3, Mr = 114.1) at 123 K [monoclinic, P2(1)/n, Z = 4, a = 10.704 (2), b = 8.187 (2), c = 4.969 (1) A, beta = 92.32 (1)o] has been determined by
L Hillered et al.
Neuroreport, 6(13), 1816-1820 (1995-09-11)
The authors used intracerebral microdialysis to harvest allantoin and parabanic acid, potential markers of in vivo oxygen radical activity, from the frontal lobe cortex of three patients in the neurointensive care unit after serious aneurysmal subarachnoid haemorrhage. Clinical events involving
H Kaur et al.
Chemico-biological interactions, 73(2-3), 235-247 (1990-01-01)
Uric acid is an end-product of purine metabolism in Man, and has been suggested to act as an antioxidant in vivo. Products of attack upon uric acid by various oxidants were measured by high performance liquid chromatography. Hypochlorous acid rapidly
T Kotani et al.
Chemical & pharmaceutical bulletin, 45(2), 297-304 (1997-02-01)
Accumulation of intracellular sorbitol, the product of glucose reduction catalyzed by aldose reductase (AR) [EC 1.1.1.21], is thought to be the main culprit in the development of diabetic complications. A series of 3-arylalkyl-2,4,5-trioxoimidazolidine-1-acetic acids was prepared and tested for inhibitory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service