Skip to Content
Merck
All Photos(1)

Key Documents

OCNICK10

Sigma-Aldrich

EDC Conjugation Kit for Iron Oxide (III) Nanoparticles

10 nm, carboxylic acid functionalized, 5 mg/mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€356.00

€356.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€356.00

About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

€356.00


Check Cart for Availability

Request a Bulk Order

form

nanoparticles

concentration

5 mg/mL

particle size

10 nm

functional group

carboxylic acid

Looking for similar products? Visit Product Comparison Guide

Application

Iron oxide nanoparticles are magnetic single crystals and can be used as magnetic resonance imaging contrast reagents for in-vivo imaging , drug delivery vehicle, hyperthermia genertor for therapy, adjuvant for vaccine, and magnetic sensing probe for in-vitro diagnostics. Ocean NanoTech′s carboxyl terminated iron oxide nanocrystals of various diameters (10 nm-30 nm) are now available in an easy-to-use kit format to enable researchers to conjugate proteins/ligands of their own choice to these iron oxide nanoparticles. The kit contains sufficient reagents & components for performing at least 5 conjugation reactions using 1mg iron oxide nanoparticles per reaction. The iron oxide nanoparticles are activated using carbodiimide and N-hydroxysuccinimide followed by conjugation to amino groups that are present on the target protein/ligand such as bovine serum albumin, streptavidin and immunoglobulin.

Legal Information

Product of Ocean Nanotech, LLC.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Stomach,large intestine,lymph node

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service