Skip to Content
Merck
All Photos(1)

Key Documents

NRS5600MWIPV7

Norell® valved 5 mm NMR tubes for intermediate pressure

frequency 600 MHz, L 7 in., medium wall

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€281.00

€281.00


Check Cart for Availability


Select a Size

Change View
1 EA
€281.00

About This Item

UNSPSC Code:
41121705
NACRES:
SB.32

€281.00


Check Cart for Availability

material

glass tube

feature

medium wall

packaging

pkg of 1 ea

manufacturer/tradename

Norell S-5-600-MW-IPV-7

parameter

10.5 bar max. pressure (150 psi) (1050 kPa, medium wall)
14 bar max. pressure (200 psi) (1400 kPa, heavy wall)
600 MHz frequency
7 bar max. pressure (100 psi) (700 kPa, thin wall)

L

7 in.

tube diam.

5 mm

Looking for similar products? Visit Product Comparison Guide

General description

These NMR tubes feature a glass/PTFE pressure valve permanently joined to a choice of available sample tubes. These pressure tubes facilitate experiments requiring conditions such as pressurized inert atmosphere blanketing, addition of reactive gaseous reagents under pressure, containment of low boiling point solvents or samples at elevated temperatures.

Legal Information

Norell is a registered trademark of Norell, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service