Skip to Content
Merck
All Photos(2)

Key Documents

N21022

Sigma-Aldrich

4-Nitrophenyl disulfide

Synonym(s):

p,p′-Dinitrodiphenyl disulfide, Bis(4-nitrophenyl) disulfide, Bis(p-nitrophenyl) disulfide, NSC 4566, NSC 677446

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€73.20

€73.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€73.20

About This Item

Linear Formula:
(O2NC6H4)2S2
CAS Number:
Molecular Weight:
308.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€73.20


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

SMILES string

[O-][N+](=O)c1ccc(SSc2ccc(cc2)[N+]([O-])=O)cc1

InChI

1S/C12H8N2O4S2/c15-13(16)9-1-5-11(6-2-9)19-20-12-7-3-10(4-8-12)14(17)18/h1-8H

InChI key

KWGZRLZJBLEVFZ-UHFFFAOYSA-N

Application

Reactant or reagent involved in:
  • Electrophilic cyclization of 2-alkynylanisoles or alkynylanilines
  • Oxidative chlorination to sulfonyl chlorides
  • Arylation with triarylbismuthanes
  • Decarboxylative cross-coupling with dialkoxybenzoic acids
  • Disulfidation of alkenes

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of p-nitrophenyl disulfide to measure reductive capacity of intact cells.
C Gitler et al.
Methods in enzymology, 251, 279-286 (1995-01-01)
Homan Kang et al.
Scientific reports, 5, 10144-10144 (2015-05-29)
Recently, preparation and screening of compound libraries remain one of the most challenging tasks in drug discovery, biomarker detection, and biomolecular profiling processes. So far, several distinct encoding/decoding methods such as chemical encoding, graphical encoding, and optical encoding have been

Articles

Markovnikov and anti-Markovnikov alkene reactivity differences are discussed, highlighting challenges and catalytic advancements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service