Skip to Content
Merck
All Photos(1)

Documents

N162

Sigma-Aldrich

R-(−)-Niguldipine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H39N3O6 · HCl
CAS Number:
Molecular Weight:
646.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

optical activity

[α]23/D −12.5°, c = 0.14 in methanol(lit.)

color

pale yellow

solubility

methanol: soluble

storage temp.

−20°C

SMILES string

Cl.COC(=O)C1=C(C)NC(C)=C([C@@H]1c2cccc(c2)[N+]([O-])=O)C(=O)OCCCN3CCC(CC3)(c4ccccc4)c5ccccc5

InChI

1S/C36H39N3O6.ClH/c1-25-31(34(40)44-3)33(27-12-10-17-30(24-27)39(42)43)32(26(2)37-25)35(41)45-23-11-20-38-21-18-36(19-22-38,28-13-6-4-7-14-28)29-15-8-5-9-16-29;/h4-10,12-17,24,33,37H,11,18-23H2,1-3H3;1H/t33-;/m1./s1

InChI key

MHOSUIMBPQVOEU-MGDILKBHSA-N

Biochem/physiol Actions

Less active enantiomer

Caution

Photosensitive. Adsorbs strongly to plastic.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Höllt et al.
Biochemical pharmacology, 43(12), 2601-2608 (1992-06-23)
The (-)-isomer of verapamil is 10-fold more potent as a calcium antagonist than the (+)-isomer. However, both enantiomers are equally effective in increasing cellular accumulation of anticancer drugs [Gruber et al., Int J Cancer 41: 224-226, 1988]. In addition to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service