Skip to Content
Merck
All Photos(1)

Key Documents

N15200

Sigma-Aldrich

4-Nitrobiphenyl

Synonym(s):

1-Nitro-4-phenylbenzene, p-Phenylnitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9NO2
CAS Number:
Molecular Weight:
199.21
Beilstein:
1910956
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

[O-][N+](=O)c1ccc(cc1)-c2ccccc2

InChI

1S/C12H9NO2/c14-13(15)12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9H

InChI key

BAJQRLZAPXASRD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Warning

OSHA-regulated carcinogen − 29 CFR Part 1910.1003

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Y Kado et al.
Mutation research, 271(3), 253-260 (1992-06-01)
Vapor-phase mutagens are potentially a major class of toxic contaminants in ambient and indoor air. These compounds are not routinely analyzed due to a lack of an established integrated methodology to quantitatively trap, extract and test the compounds in a
Laboratory decontamination and destruction of carcinogens in laboratory wastes: some aromatic amines and 4-nitrobiphenyl.
M Castegnaro et al.
IARC scientific publications, (64)(64), 1-85 (1985-01-01)
S Swaminathan et al.
Mutation research, 172(1), 37-45 (1986-10-01)
Xanthine oxidase catalyzed mutagenicity of 4-nitrobiphenyl (NBP), a dog-bladder carcinogen, was tested in Ames assay using Salmonella typhimurium TA98 strains. NBP was active as a mutagen in the parent strain TA98 which is proficient in nitroreductase, while it was inactive
S J Culp et al.
Mutation research, 378(1-2), 97-112 (1997-08-01)
4-Aminobiphenyl (ABP) is a recognized human bladder carcinogen, whose presence in cigarette smoke results in DNA adduct formation in the human urothelium. Since preliminary studies indicated that even higher levels of ABP-DNA adducts may be present in human peripheral lung
K Tatsumi et al.
Cancer research, 46(3), 1089-1093 (1986-03-01)
Reductive metabolism of aromatic nitro compounds was examined with rabbit liver preparations. Under anaerobic conditions, carcinogenic 2-nitrofluorene, 4-nitrobiphenyl, and 1-nitro-naphthalene were reduced to the corresponding hydroxylamines and amines, whereas the carcinogenic 1-nitropyrene was reduced only to the corresponding amine by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service