Skip to Content
Merck
All Photos(1)

Documents

MT51344778

Mettler-Toledo ISE standard solution

Sodium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51191600
NACRES:
NB.79

form

liquid

feature

1000 mg/L Concentration

manufacturer/tradename

Mettler-Toledo 51344778

volume

500 mL

Looking for similar products? Visit Product Comparison Guide

General description

ISE standad for ion-selective electrodes

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service