Skip to Content
Merck
All Photos(1)

Key Documents

MT51302153

Mettler-Toledo conductivity standard

Conductivity: 84 μs/cm, volume 250 mL

Synonym(s):

conductivity calibration standard

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€46.80

€46.80


Check Cart for Availability


Select a Size

Change View
250 ML
€46.80

About This Item

UNSPSC Code:
51191600
NACRES:
NB.79

€46.80


Check Cart for Availability

form

liquid

manufacturer/tradename

Mettler-Toledo 51302153

volume

250 mL

Looking for similar products? Visit Product Comparison Guide

Application

The Mettler-Toledo conductivity standard is ideal for accurate conductivity measurements.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service