M87800
1-Morpholinocyclohexene
98%
Synonym(s):
4-(1-Cyclohexen-1-yl)morpholine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H17NO
CAS Number:
Molecular Weight:
167.25
Beilstein:
118696
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.514 (lit.)
bp
118-120 °C/10 mmHg (lit.)
density
0.995 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
C1CCC(=CC1)N2CCOCC2
InChI
1S/C10H17NO/c1-2-4-10(5-3-1)11-6-8-12-9-7-11/h4H,1-3,5-9H2
InChI key
IIQFBBQJYPGOHJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
154.4 °F - closed cup
Flash Point(C)
68 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service