Skip to Content
Merck
All Photos(1)

Documents

M72404

Sigma-Aldrich

2-Methylpiperazine

95%

Synonym(s):

(±)-2-Methylpiperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2
CAS Number:
Molecular Weight:
100.16
Beilstein:
383566
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

bp

155 °C/763 mmHg (lit.)

mp

61-63 °C (lit.)

SMILES string

CC1CNCCN1

InChI

1S/C5H12N2/c1-5-4-6-2-3-7-5/h5-7H,2-4H2,1H3

InChI key

JOMNTHCQHJPVAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matteo Arioli et al.
Polymers, 12(6) (2020-06-25)
This paper aims at elucidating the degradation mechanism of linear polyamidoamines (PAAs) in water. PAAs are synthesized by the aza-Michael polyaddition of prim-monoamines or bis-sec-amines with bisacrylamides. Many PAAs are water-soluble and have potential for biotechnological applications and as flame-retardants.
Sin Ho Kweon et al.
Archives of pharmacal research, 38(11), 2020-2028 (2015-05-10)
Sesquiterpene lactone compounds have received considerable attention in pharmacological research due to their therapeutic effects including anti-cancer and anti-inflammatory activities. In this report, we investigated the effect of arsantin, a sesquiterpene lactone compound present in Artemisia santolina, on cellular differentiation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service