Skip to Content
Merck
All Photos(1)

Key Documents

M48309

Sigma-Aldrich

4-Methyl-3-heptanol, mixture of isomers

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(CH3)CH(OH)C2H5
CAS Number:
Molecular Weight:
130.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99%

refractive index

n20/D 1.43 (lit.)

bp

98-99 °C/75 mmHg (lit.)

density

0.827 g/mL at 25 °C (lit.)

SMILES string

CCCC(C)C(O)CC

InChI

1S/C8H18O/c1-4-6-7(3)8(9)5-2/h7-9H,4-6H2,1-3H3

InChI key

BKQICAFAUMRYLZ-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen T Ferguson et al.
The Journal of experimental biology, 223(Pt 2) (2020-01-05)
In eusocial ants, aggressive behaviors require the ability to discriminate between chemical signatures such as cuticular hydrocarbons that distinguish nestmate friends from non-nestmate foes. It has been suggested that a mismatch between a chemical signature (label) and the internal, neuronal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service