M46055
3-Methylene-2-norbornanone
97%
Synonym(s):
3-methylene-bicyclo[2,2,1]heptan-2-one
About This Item
Recommended Products
Assay
97%
refractive index
n20/D 1.493 (lit.)
bp
69-71 °C/11 mmHg (lit.)
density
0.997 g/mL at 25 °C (lit.)
SMILES string
C=C1[C@@H]2CC[C@@H](C2)C1=O
InChI
1S/C8H10O/c1-5-6-2-3-7(4-6)8(5)9/h6-7H,1-4H2/t6-,7+/m1/s1
InChI key
FNOOZJAPZFHNCW-RQJHMYQMSA-N
Application
- 3-(2-(4-methoxyphenyl)-2-oxoethyl)bicyclo[2.2.1]heptan-2-one by reacting with 4-anisaldehyde in the presence of tetrabutylammonium decatungstate (TBADT) photocatalyst.
- 2-Aryl-3-methylene-endo-norbornanols by treating with corresponding aryl lithium reagents.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
131.0 °F - closed cup
Flash Point(C)
55 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service