Skip to Content
Merck
All Photos(1)

Key Documents

M38583

Sigma-Aldrich

(R)-(+)-3-Methylcyclohexanone

98%

Synonym(s):

(+)-(3R)-3-Methylcyclohexanone, (+)-3(R)-Methylcyclohexanone, (+)-3-Methyl-1-cyclohexanone, (3R)-(+)-3-Methylcyclohexanone, (3R)-3-Methylcyclohexanone, (3R)-Methylcyclohexanone, (R)-3-Methylcyclohexanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H9(=O)
CAS Number:
Molecular Weight:
112.17
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

optical activity

[α]24/D +13.5°, neat

optical purity

ee: 99% (GLC)

refractive index

n20/D 1.446 (lit.)

bp

168-169 °C (lit.)

density

0.916 g/mL at 25 °C (lit.)

SMILES string

C[C@@H]1CCCC(=O)C1

InChI

1S/C7H12O/c1-6-3-2-4-7(8)5-6/h6H,2-5H2,1H3/t6-/m1/s1

InChI key

UJBOOUHRTQVGRU-ZCFIWIBFSA-N

Application

(R)-(+)-3-Methylcyclohexanone may be used in the preparation of (R)-5-methyl-2-(1-methylethylidene-d6)-cyclohexanone [(R)-(+)-pulegone-d6].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantiospecific Adsorption of (R)-3-Methylcyclohexanone on Naturally Chiral Cu (531) R&S Surfaces
Huang, Ye, and Andrew J. Gellman
Catalysis Letters, 125-3.4, 177-182 (2008)
Polarized Raman optical activity and the bond polarizability model
Barron, L. D., J. R. Escribano, and J. F. Torrance
Molecular Physics, 57.3, 653-660 (1986)
The metabolism of the abortifacient terpene,(R)-(+)-pulegone, to a proximate toxin, menthofuran.
Gordon WP, et al.
Drug Metabolism and Disposition, 15(5), 589-594 (1987)
Linear and nonlinear circular dichroism of R-(+)-3-methylcyclopentanone
Li, R., et al.
J. Chem. Phys. , 125.14, 144304-144304 (2006)
Enantiospecific orientation of R?3-methylcyclohexanone on the chiral Cu (643) R/S surfaces
Horvath, Joshua D., Layton Baker, and Andrew J. Gellman
The Journal of Physical Chemistry C, 112.20, 7637-7343 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service