Skip to Content
Merck
All Photos(1)

Key Documents

M24809

Sigma-Aldrich

4′-Methoxypropiophenone

≥99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€69.10

€69.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€69.10

About This Item

Linear Formula:
CH3OC6H4COC2H5
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€69.10


Check Cart for Availability

Request a Bulk Order

Assay

≥99%

refractive index

n20/D 1.5465 (lit.)

bp

273-275 °C (lit.)

mp

27-29 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

SMILES string

CCC(=O)c1ccc(OC)cc1

InChI

1S/C10H12O2/c1-3-10(11)8-4-6-9(12-2)7-5-8/h4-7H,3H2,1-2H3

InChI key

ZJVAWPKTWVFKHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mence Najdoska-Bogdanov et al.
Natural product communications, 11(3), 423-429 (2016-05-14)
Two extraction methods for subsequent gas chromatographic (GC) determination of volatiles from freshly harvested and aged fennel fruit samples (Foeniculum vulgare Mill.,ssp. vulgare var. dulce) have been compared. Hydrodistillation followed by GC-FID and GC-MS analysis was used as a standard
Jiang Yan et al.
Science advances, 6(45) (2020-11-08)
Phenol is an important commodity chemical in the industry, which is currently produced using fossil feedstocks. Here, we report a strategy to produce phenol from lignin by directly deconstructing Csp2-Csp3 and C-O bonds under mild conditions. It was found that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service