M2014
5-Methoxy-3-(2-nitrovinyl)indole
Synonym(s):
5-Methoxy-3-(2′-nitroethenyl)indole, NSC 88875
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H10N2O3
CAS Number:
Molecular Weight:
218.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
SMILES string
COc1ccc2[nH]cc(\C=C\N(=O)=O)c2c1
InChI
1S/C11H10N2O3/c1-16-9-2-3-11-10(6-9)8(7-12-11)4-5-13(14)15/h2-7,12H,1H3/b5-4+
InChI key
NAIAZIKEFZGLDL-SNAWJCMRSA-N
Application
Reactant for preparation of:
- Inhibitors of mitogen activated protein kinase-activated protein kinase 2 (MK-2)
- Indole lipoic acid derivatives as antioxidants effective against lipid peroxidation
- Indole ethylamine derivatives as melatonin analogs
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service