M14870
5-Methoxygramine
99%
Synonym(s):
3-(Dimethylaminomethyl)-5-methoxyindole, NSC 88885
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H16N2O
CAS Number:
Molecular Weight:
204.27
Beilstein:
170533
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
mp
123-126 °C (lit.)
SMILES string
COc1ccc2[nH]cc(CN(C)C)c2c1
InChI
1S/C12H16N2O/c1-14(2)8-9-7-13-12-5-4-10(15-3)6-11(9)12/h4-7,13H,8H2,1-3H3
InChI key
GOERTRUXQHDLHC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Reactant for preparation of dopamine D2 receptor antagonists
- Reactant for asymmetric preparation of ethenyl(tetrahydro)carbazoledicarboxylates via gold(I)-catalyzed intramolecular Friedel-Crafts allylic alkylation reaction
- Reactant for preparation of N-substituted 3-aryl-8-azabicyclo[3.2.1]octan-3-ols as dopamine D2-like receptor ligands
- Reactant for preparation of substituted N-trimethoxybenzoyl indoles, indolines, and a tetrahydroquinoline via N-aroylation with trimethoxybenzoyl chloride or anhydride as antitubulin agents
- Reactant for preparation of indolylmethanesulfonamide and its methoxy derivatives
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
B M Clancy et al.
The Journal of pharmacology and experimental therapeutics, 242(1), 108-114 (1987-07-01)
The kinetics of agonist and antagonist interactions with the 5-hydroxytryptamine2 receptor were studied in the isolated rabbit aorta by following the antagonist-induced decrease in the steady-state response to an agonist. A model describing the competitive drug-receptor interactions was fitted to
Mark G Devlin et al.
European journal of pharmacology, 498(1-3), 59-69 (2004-09-15)
The effect of ligand pretreatment on human 5-hydroxytryptamine2C (5-HT2C) receptors was examined in CHO cells expressing high (CHO-1C7; 67+/-3 pmol/mg) or low (CHO-1C19; 72+/-10 fmol/mg) levels of the receptor. Seventy-two hours pretreatment of CHO-1C7 cells with various ligands did not
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service