Skip to Content
Merck
All Photos(1)

Documents

M1021

Sigma-Aldrich

1-Methyl-4-phenyl-1,2,3,6-tetrahydropyridine

Synonym(s):

MPTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N
CAS Number:
Molecular Weight:
173.25
EC Number:
MDL number:
UNSPSC Code:
12352100

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

bp

128-132 °C/12 mmHg (lit.)

SMILES string

CN1CCC(=CC1)c2ccccc2

InChI

1S/C12H15N/c1-13-9-7-12(8-10-13)11-5-3-2-4-6-11/h2-7H,8-10H2,1H3

InChI key

PLRACCBDVIHHLZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dopaminergic neurotoxin.

Warning

Reportedly causes a severe and irreversible Parkinsonism condition in humans and monkeys. New Engl. J. Med. 1983, 309, 310. Science 1983, 219, 979. ibid. 1984, 224, 1083, 1451. ibid. 1984, 225, 529. J. Med. Chem. 1985, 28, 997.

Pictograms

Health hazardSkull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cécile Moro et al.
BMC neuroscience, 14, 40-40 (2013-03-28)
We have shown previously that near-infrared light (NIr) treatment or photobiomodulation neuroprotects dopaminergic cells in substantia nigra pars compacta (SNc) from degeneration induced by 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) in Balb/c albino mice, a well-known model for Parkinson's disease. The present study explores
Some features of the nigrostriatal dopaminergic neurotoxin 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) in the mouse.
R E Heikkila et al.
Molecular and chemical neuropathology, 10(3), 171-183 (1989-06-01)
Sang-Bing Ong et al.
Thrombosis and haemostasis, 113(3), 513-521 (2014-09-26)
The mechanism through which the protein kinase Akt (also called PKB), protects the heart against acute ischaemia-reperfusion injury (IRI) is not clear. Here, we investigate whether Akt mediates its cardioprotective effect by modulating mitochondrial morphology. Transfection of HL-1 cardiac cells
Serge Przedborski et al.
Annals of the New York Academy of Sciences, 991, 189-198 (2003-07-09)
Experimental models of dopaminergic neurodegeneration play a critical role in our quest to elucidate the cause of Parkinson's disease (PD). Despite the recent development of "genetic models" that have followed upon the discovery of mutations causing rare forms of familial
Rui D S Prediger et al.
Current pharmaceutical design, 17(5), 489-507 (2011-03-08)
Parkinson's disease (PD) is the second most common neurodegenerative disorder affecting approximately 1% of the population older than 60 years. Classically, PD is considered to be a motor system disease and its diagnosis is based on the presence of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service