Skip to Content
Merck
All Photos(1)

Key Documents

LGFG40K

Sigma-Aldrich

Green Fluorescent PLGA microspheres

40 μm average diameter

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€558.00

€558.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€558.00

About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

€558.00


Check Cart for Availability

Request a Bulk Order

description

Excitation/Emission: 460/500 nm
Excitation/Emission: 460/500 nm

form

pellets

average diameter

40 μm

storage temp.

-10 to -25°C

Looking for similar products? Visit Product Comparison Guide

Application

Fluorescent PLGA particles for imaging and cell tracking in drug discovery. Particles can be used for fluorscent assays and drug delivery.

Legal Information

Product of Phosphorex, Inc.Degradex(RM) is a product of Phosphorex, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric antioxidants show promise in drug delivery with superior bioavailability and antioxidant potential.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service