Skip to Content
Merck
All Photos(1)

Key Documents

L512117

Sigma-Aldrich

BCzVB

AldrichCPR

Synonym(s):

1,4-Bis[2-(3-N-ethylcarbazoryl)-vinyl]benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H32N2
CAS Number:
Molecular Weight:
516.67
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

CCN1C2=C(C=CC=C2)C3=C1C=CC(/C=C/C4=CC=C(/C=C/C5=CC(C(C=CC=C6)=C6N7CC)=C7C=C5)C=C4)=C3

InChI

1S/C38H32N2/c1-3-39-35-11-7-5-9-31(35)33-25-29(21-23-37(33)39)19-17-27-13-15-28(16-14-27)18-20-30-22-24-38-34(26-30)32-10-6-8-12-36(32)40(38)4-2/h5-26H,3-4H2,1-2H3/b19-17+,20-18+

InChI key

JQXNAJZMEBHUMC-XPWSMXQVSA-N

Application

Blue-emitting material for full color displays. It may be used as a blue emitter for a double emitting layer in white organic light emitting diodes (WOLEDs).

1,4-Bis[2-(3-N-ethylcarbazoryl)-vinyl]benzene (BCzVB) is popularly used on highly efficient pure blue electroluminescence OLEDs [7]

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient pure blue electroluminescence from 1,4-bis[2-(3-N-ethylcarbazoryl)vinyl]benzene
Wu et al.
Applied Physics Letters, 83, 5077-5077 (2003)
Enhancing color purity and efficiency of white organic light-emitting diodes using a double-emitting layer.
Lin Hua-Ping, et al.
Semiconductor Science and Technology null
Enhanced color stability and improved performance in white organic light-emitting devices by utilizing a double-graded structure
Lin Hua-Ping, et al.
Synt. Metals null
Enhancing color purity and efficiency of white organic light-emitting diodes using a double-emitting layer.
Lin Hua-Ping, et al.
Semiconductor Science and Technology, 105004 null
Pure blue organic light-emitting devices based on 2, 5-diphenyl-1, 4-distyrylbenzene with two trans-double bonds.
Cheng , et al.
Semiconductor Science and Technology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service