Skip to Content
Merck
All Photos(1)

Key Documents

L040082

Sigma-Aldrich

Cap A

Sign Into View Organizational & Contract Pricing

Select a Size

45 L
€2,250.00

€2,250.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
45 L
€2,250.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.21

€2,250.00


Check Cart for Availability

Request a Bulk Order

type

for DNA synthesis

Quality Level

product line

Proligo Reagents

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Acetic Anhydride / Tetrahydrofurane 9.1/90.9 (v/v)

Other Notes

Cap A solution, containing acetic anhydride and tetrahydrofuran (THF), is used after the phosphoramidite reaction/coupling step. This solution aborts chains bearing unreacted 5′hydroxyl groups due to failure of reaction with activated phosphoramidite (typically, 1 to 2% of growing oligonucleotide chains in each synthesis cycle). These unreacted 5′hydroxyl groups are capped with an acetyl group and thus rendered unreactive for subsequent synthesis steps.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

6.1 °F

Flash Point(C)

-14.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service