Skip to Content
Merck
All Photos(1)

Key Documents

L032080

Sigma-Aldrich

DCI Activator

0.25 M, configured for PerkinElmer, configured for Polygen

Synonym(s):

DCI activator solution

Sign Into View Organizational & Contract Pricing

Select a Size

6 X 200 ML
€294.00

€294.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
6 X 200 ML
€294.00

About This Item

UNSPSC Code:
41116105
NACRES:
NA.21

€294.00


Check Cart for Availability

Request a Bulk Order

type

for DNA synthesis
for DNA synthesis

product line

Proligo Reagents

storage condition

dry at room temperature

concentration

29-32 g/L (DCI content, titration)

impurities

≤30 ppm water content (Karl Fischer)
≤30 ppm water content (Karl Fischer)

color

clear
clear

compatibility

configured for PerkinElmer
configured for Polygen

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Dicyanoimidazole / Acetonitrile 3/100 (w/v)

Other Notes

Activator solution, containing 4,5-dicyanoimidazole (DCI) in acetonitrile, are mixed with solutions of phosphoramidites during the extension step.
The activator reacts with the amidite group to form a highly reactive intermediate. The intermediate then forms an internucleotide bond with the detritylated 5′hydroxyl group of the growing oligonucleotide chain.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F

Flash Point(C)

5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service