Skip to Content
Merck
All Photos(1)

Key Documents

JKA10010

Sigma-Aldrich

8arm-PEG10K-Succinimidyl Glutarate

tripentaerythritol core, average Mn 10,000

Synonym(s):

8arm-PEG-Succinimidyl Glutarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
R(O(CH2CH2O)nCOCH2CH2CH2COOC4H4O2N)8
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 10,000

reaction suitability

reactivity: amine reactive

polymer architecture

shape: 8-arm
functionality: homofunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

8arm PEG Succinimidyl Glutarate. Crosslinks into hydrogels. Cleavable PEG linker. The ester linker between PEG and NHS ester facilitates the formation of degradable hydrogel. 8ARM(TP)-PEG with tripentaerythritol core has a lower polydispersity and higher molecular weight accuracy compared with the generic 8ARM-PEG with a hexaglycerin core.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service