Skip to Content
Merck
All Photos(1)

Key Documents

I3877

Sigma-Aldrich

Chlorpropham

~95%, crystalline

Synonym(s):

Chloro-ICP, Chloropropham, Isopropyl N-(3-chlorophenyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H12NO2Cl
CAS Number:
Molecular Weight:
213.66
Beilstein:
2211397
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

~95%

form

crystalline

color

light tan

SMILES string

CC(C)OC(=O)Nc1cccc(Cl)c1

InChI

1S/C10H12ClNO2/c1-7(2)14-10(13)12-9-5-3-4-8(11)6-9/h3-7H,1-2H3,(H,12,13)

InChI key

CWJSHJJYOPWUGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A mitotic poison; inhibits plant metabolism.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - STOT RE 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multi-residue method for determination of pesticides residues in potatoes by GC-NPD.
O Pigeon et al.
Communications in agricultural and applied biological sciences, 70(4), 1083-1086 (2006-04-25)
Xia Ge et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 81(1), 397-403 (2011-07-16)
An inclusion complex between the agrochemical chloropropham (CIPC) and β-cyclodextrin (β-CD) was prepared. A 2:1 host-guest stoichiometry was conformed by elemental analysis. From the phase solubility studies, the calculated stepwise stability constants were K(1)=224.6L/mol and K(2)=939.2L/mol, respectively. FT-IR, thermoanalysis and
Margaret J Smith et al.
Journal of hazardous materials, 246-247, 154-162 (2013-01-09)
DFT calculations were performed on the thermal reactions of chlorpropham 1, a carbamate pesticide and plant growth regulator frequently used in the storage of potatoes. At the conditions normally used in applying 1 (injection of a methanolic solution of 1
T Fujitani et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 39(3), 253-259 (2001-03-30)
Male F344 rats were given 0 or 3% chlorpropham in the diet and at 2, 4, 6, 8 or 13 weeks of administration, five rats in each group were examined for hematology, plasma clinical chemistry and pathology. Marked splenomegaly and
Ona Sakaliene et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 44(1), 1-6 (2008-12-18)
Chlorpropham (isopropyl 3-chlorocarbanilate) is a pesticide used to control sprouting of potatoes during long-term storage. The objective of the present study was to establish the total chlorpropham residue balance (residues in unwashed and washed whole tubers, peeled tubers, peels, boiled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service