I2606
3-Indazolinone
97%
Synonym(s):
3-Hydroxy-1H-indazole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H6N2O
CAS Number:
Molecular Weight:
134.14
Beilstein:
3733
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
250-252 °C (lit.)
SMILES string
O=C1NNc2ccccc12
InChI
1S/C7H6N2O/c10-7-5-3-1-2-4-6(5)8-9-7/h1-4H,(H2,8,9,10)
InChI key
SWEICGMKXPNXNU-UHFFFAOYSA-N
Gene Information
rat ... Alox5(25290)
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Pedro González-Naranjo et al.
Bioorganic & medicinal chemistry, 28(19), 115672-115672 (2020-09-12)
Synthesis and pharmacological evaluation of a new series of cannabinoid receptor antagonists of indazole ether derivatives have been performed. Pharmacological evaluation includes radioligand binding assays with [3H]-CP55940 for CB1 and CB2 receptors and functional activity for cannabinoid receptors on isolated
S D Wyrick et al.
Journal of medicinal chemistry, 27(6), 768-772 (1984-06-01)
The apparent benefit of limiting serum cholesterol and triglyceride levels either by dietary restriction or drug therapy has prompted work in our laboratories toward development of a suitable antihyperlipidemic agent. We have demonstrated the antihyperlipidemic activity of a series of
G E Demas et al.
Molecular medicine (Cambridge, Mass.), 3(9), 610-616 (1997-11-05)
Mice with targeted disruption of the gene for the neuronal isoform of nitric oxide synthase (nNOS) display exaggerated aggression. Behavioral studies of mice with targeted gene deletions suffer from the criticism that the gene product is missing not only during
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service