Skip to Content
Merck
All Photos(1)

Key Documents

I2215

Sigma-Aldrich

5-Indanol

99%

Synonym(s):

5-Hydroxyindan

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€114.00

€114.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€114.00

About This Item

Empirical Formula (Hill Notation):
C9H10O
CAS Number:
Molecular Weight:
134.18
Beilstein:
1936314
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€114.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

bp

255 °C (lit.)

mp

51-53 °C (lit.)

SMILES string

Oc1ccc2CCCc2c1

InChI

1S/C9H10O/c10-9-5-4-7-2-1-3-8(7)6-9/h4-6,10H,1-3H2

InChI key

PEHSSTUGJUBZBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Dermal

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service