Key Documents
HAM61401-01
Concorde convertible tip hand probe, for Hamilton® Microlab® 600 pipettor
pack of 1 ea
Select a Size
1 EA
€300.00
€300.00
Check Cart for Availability
Select a Size
Change View
1 EA
€300.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
€300.00
Check Cart for Availability
feature
Probe receptacle electrical connection
packaging
pack of 1 ea
cable L
1200 mm
gauge
41626
General description
The Concorde Convertible Tip Hand Probe is designed to work with the Microlab 600. It is shipped standard with the single syringe dispenser, dual syringe diluter and the continuous dispenser.
This probe is most commonly used with 12 or 18 gauge tubing but may be used with tubing as large as 1/8 inch for viscous liquids. A red/green LED incorporated into the hand probe alerts the user when the instrument is ready to trigger.
For comfortable operation the probe tip can be oriented at either 0 or 15 degrees. Using the Luer Lock Conversion Kit the tip can also be converted into a Luer Lock termination that can be used with standard Luer Lock needles.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)