Key Documents
HAM201400
Hamilton® Gastight® syringe, general use, 1000 series
model, 1025, AccuDil (AD) end, volume 25 mL
Select a Size
1 EA
€313.00
€313.00
Check Cart for Availability
Select a Size
Change View
1 EA
€313.00
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
€313.00
Check Cart for Availability
material
Borosilicate glass/PTFE
Gastight syringe
feature
autoclavable: no
model 1025, AccuDil (AD) end
manufacturer/tradename
Hamilton
parameter
20-50 °C temp. range (68-122 °F)
200 psig pressure
volume
25 mL
Related Categories
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)