All Photos(1)
About This Item
Linear Formula:
HOCH2(CF2)3CH2OH
CAS Number:
Molecular Weight:
212.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
98%
bp
111.5 °C/10 mmHg (lit.)
mp
78-81 °C (lit.)
SMILES string
OCC(F)(F)C(F)(F)C(F)(F)CO
InChI
1S/C5H6F6O2/c6-3(7,1-12)5(10,11)4(8,9)2-13/h12-13H,1-2H2
InChI key
IELVMUPSWDZWSD-UHFFFAOYSA-N
Application
Used for the preparation of cyclic and acyclic polyfluorosiloxanes as precursors to per- and polyfluoroethers.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Inorganic Chemistry, 33, 415-415 (1994)
Inorganic Chemistry, 5463-5463 null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service